missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H2/ICOSLG Antibody (599829) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
216.00 € - 497.00 €
Specifications
| Antigen | B7-H2/ICOSLG |
|---|---|
| Clone | 599829 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18078274
|
Novus Biologicals
MAB1581-SP |
25 μg |
216.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633754
|
Novus Biologicals
MAB1581 |
100 μg |
497.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| B7-H2/ICOSLG | |
| 1.0 mg/mL | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Mouse | |
| B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| ICOSLG | |
| IgG1 | |
| Protein A or G purified |
| 599829 | |
| Flow Cytometry, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with No Preservative | |
| 23308 | |
| rmB7-H2 aa 47-279 | |
| Primary | |
| Store at -20C in the dark. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title