missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H2/ICOSLG Antibody (MM0103-19G23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12047
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications
| B7-H2/ICOSLG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Flow Cytometry, Block/Neutralize | |
| O75144 | |
| ICOSLG | |
| Human recombinant B7-H2/ICOSLG (extracellular domain) | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 23308 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Neutralization | |
| MM0103-19G23 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction