missing translation for 'onlineSavingsMsg'
Learn More

Bad Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

302.00 € - 498.00 €

Specifications

Antigen Bad
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18616075
View Documents
Novus Biologicals
NBP2-38947-25ul
25 μL
302.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18158128
View Documents
Novus Biologicals
NBP2-38947
0.1 mL
498.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bad Polyclonal specifically detects Bad in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Bad
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q92934
572
This antibody was developed against a recombinant protein corresponding to amino acids: NLWAAQRYGRELRRMSDEFVDSFKKGLPRPKSAGTATQMRQSSSWTRVFQSWWDRNLGR
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Rabbit
Angiogenesis, Apoptosis, Cancer, Death Receptor Signaling Pathway, GPCR, Neuroscience, Tumor Suppressors
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter
BAD
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.