missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAD-LAMP/LAMP5 Antibody (LAMP5/7642), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23810-20ug
This item is not returnable.
View return policy
Description
BAD-LAMP/LAMP5 Monoclonal antibody specifically detects BAD-LAMP/LAMP5 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| BAD-LAMP/LAMP5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| BADLAMP, C20orf103, chromosome 20 open reading frame 103, dJ1119D9.3, LAMP family protein C20orf103, LAMP5, LAMP-5, lysosomal-associated membrane protein family, member 5, UNC-43 | |
| Recombinant full-length human BAD-LAMP/LAMP5 protein | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| LAMP5/7642 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 24141 | |
| Store at 4°C. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction