missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAD-LAMP/LAMP5 Antibody (LAMP5/7642), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | BAD-LAMP/LAMP5 |
|---|---|
| Clone | LAMP5/7642 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665584
|
Novus Biologicals
NBP3-23810-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691785
|
Novus Biologicals
NBP3-23810-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAD-LAMP/LAMP5 Monoclonal antibody specifically detects BAD-LAMP/LAMP5 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| BAD-LAMP/LAMP5 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| BADLAMP, C20orf103, chromosome 20 open reading frame 103, dJ1119D9.3, LAMP family protein C20orf103, LAMP5, LAMP-5, lysosomal-associated membrane protein family, member 5, UNC-43 | |
| Recombinant full-length human BAD-LAMP/LAMP5 protein | |
| Primary | |
| Store at 4°C. |
| LAMP5/7642 | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 24141 | |
| IgG2 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title