missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BAFFR/TNFRSF13C Antibody (5A9B6), Novus Biologicals™

Product Code. 30033407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30033407 0.1 mL 0.1mL
30031636 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30033407 Supplier Novus Biologicals™ Supplier No. NBP3271270.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human,Rat samples. It is validated for Flow Cytometry,ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry, ELISA, Western Blot
Classification Monoclonal
Clone 5A9B6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Formulation PBS
Gene Alias BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C
Host Species Mouse
Immunogen Purified recombinant fragment of human BAFFR/TNFRSF13C (AA: extra 1-78) expressed in E. Coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.