missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG2 Mouse anti-Human, Clone: 2.90E+10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59476-100UL
This item is not returnable.
View return policy
Description
BAG2 Monoclonal specifically detects BAG2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BAG2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1074, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| BAG family molecular chaperone regulator 2, BAG-2, BAG-family molecular chaperone regulator-2, Bcl-2-associated athanogene 2, BCL2-associated athanogene 2, dJ417I1.2, dJ417I1.2 (BAG-family molecular chaperone regulator 2), KIAA0576, MGC149462 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 29000000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| BAG2 | |
| Recombinant human BAG2 (1-211aa) purified from E. coli | |
| 100 μL | |
| Apoptosis | |
| 9532 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction