missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG2 Mouse anti-Human, Clone: 2.90E+10, Novus Biologicals™
Mouse Monoclonal Antibody
319.00 € - 497.00 €
Specifications
| Antigen | BAG2 |
|---|---|
| Clone | 29000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1074, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18073264
|
Novus Biologicals
NBP2-59476-50UL |
50 μL |
319.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284123
|
Novus Biologicals
NBP2-59476-100UL |
100 μL |
497.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAG2 Monoclonal specifically detects BAG2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| BAG2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 9532 | |
| Recombinant human BAG2 (1-211aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 29000000000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1074, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAG family molecular chaperone regulator 2, BAG-2, BAG-family molecular chaperone regulator-2, Bcl-2-associated athanogene 2, BCL2-associated athanogene 2, dJ417I1.2, dJ417I1.2 (BAG-family molecular chaperone regulator 2), KIAA0576, MGC149462 | |
| BAG2 | |
| IgG2a κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title