missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BAG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30852-100ug
This item is not returnable.
View return policy
Description
BAG5 Polyclonal antibody specifically detects BAG5 in Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| BAG5 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, Immunohistochemistry-Paraffin | |
| BAG family molecular chaperone regulator 5, BAG-5, BAG-family molecular chaperone regulator-5, Bcl-2-associated athanogene 5, BCL2-associated athanogene 5, KIAA0873 | |
| Synthetic peptide derived from the human BAG5 protein. | |
| 100 μg | |
| Apoptosis | |
| 9529 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction