missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BAG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | BAG5 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067239
|
Novus Biologicals™
NBP3-30852-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067240
|
Novus Biologicals™
NBP3-30852-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAG5 Polyclonal antibody specifically detects BAG5 in Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
| BAG5 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| Phosphate buffered saline | |
| 9529 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| BAG family molecular chaperone regulator 5, BAG-5, BAG-family molecular chaperone regulator-5, Bcl-2-associated athanogene 5, BCL2-associated athanogene 5, KIAA0873 | |
| Synthetic peptide derived from the human BAG5 protein. | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title