Learn More
BAHCC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30046-1-AP-150UL
Description
The BAHCC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAHCC1. This antibody recognizes human, mouse, rat antigen. The BAHCC1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| BAHCC1 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| BAHCC1, BAHD2, FLJ23058, KIAA1447 | |
| 150 μL | |
| 57597 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |