missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAHCC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | BAHCC1 |
|---|---|
| Concentration | 500 μg/mL |
| Applications | Immunofluorescence, Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020536
|
Proteintech
30046-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095195
|
Proteintech
30046-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BAHCC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAHCC1. This antibody recognizes human, mouse, rat antigen. The BAHCC1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| BAHCC1 | |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| Rabbit | |
| 57597 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 500 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| BAHCC1, BAHD2, FLJ23058, KIAA1447 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title