Learn More
BAHCC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30046-1-AP-20UL
 View more versions of this product
Product Code. 18095195
  • 145.00 € / 20µL
Description
The BAHCC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAHCC1. This antibody recognizes human, mouse, rat antigen. The BAHCC1 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
BAHCC1
500 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Immunofluorescence, Immunohistochemistry, ELISA
BAHCC1, BAHD2, FLJ23058, KIAA1447
20 μL
57597
Store at -20°C. Stable for one year after shipment.