missing translation for 'onlineSavingsMsg'
Learn More

Bamethan Sulfate, Mikromol™

Product Code. 30050711
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050711 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050711 Supplier Mikromol Supplier No. MM1822.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bamethan Sulfate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bamethan Sulfate
CAS 5716-20-1
Analyte or Component Names Bamethan Sulfate
Shipping Condition Room Temperature
Molecular Formula 2 C12 H19 N O2 . H2 O4 S
Certifications/Compliance ISO 17025
Synonym Benzenemethanol, α-[(butylamino)methyl]-4-hydroxy-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(butylamino)methyl]-p-hydroxy-, sulfate (2:1) (salt) (8CI), (±)-1-(4-Hydroxyphenyl)-1-hydroxy-2-butylaminoethane sulfate, 1-(4-Hydroxyphenyl)-2-butylaminoethanol-sulfate, 2-n-Butylamino-1-(p-hydroxyphenyl)ethanol sulfate, Bametan sulfate, Bamethan sulfate, Bascurat, Bupatol, Butedrin, Butibatol, Butylnorsympatol, Cyclate, Eclern, Garmian, Periphetol, Rotesar, Vasculat, Vasculit, Vaskulat, α-[(Butylamino)methyl]-p-hydroxybenzyl alcohol sulfate (2:1), Benzenemethanol, α-[(butylamino)methyl]-4-hydroxy-, sulfate (2:1), Bamethan Sulfate
InChI Formula InChI=1S/2C12H19NO2.H2O4S/c2*1-2-3-8-13-9-12(15)10-4-6-11(14)7-5-10;1-5(2,3)4/h2*4-7,12-15H,2-3,8-9H2,1H3;(H2,1,2,3,4)
SMILES CCCCNCC(O)c1ccc(O)cc1.CCCCNCC(O)c2ccc(O)cc2.OS(=O)(=O)O
IUPAC Name 4-[2-(butylamino)-1-hydroxyethyl]phenol;sulfuric acid
Molecular Weight (g/mol) 516.648
Quantity 100 mg
Formula Weight 516.2505 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.