missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP28 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | BAP28 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629830
|
Novus Biologicals
NBP2-92105-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698211
|
Novus Biologicals
NBP2-92105-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAP28 Polyclonal antibody specifically detects BAP28 in Human samples. It is validated for Western BlotSpecifications
| BAP28 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS with 50% glycerol, pH7.3. | |
| 55127 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BAP28HEAT repeat-containing protein 1, EC 4.1.1.31, EC 6.3.4.4, FLJ10359, HEAT repeat containing 1, MGC72083, Protein BAP28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human HEATR1 (NP_060542.4). MTSLAQQLQRLALPQSDASLLSRDEVASLLFDPKEAATIDRDTAFAIGCTGLEELLGIDPSFEQFEAPLFSQLAKTLERSVQTKAVNKQLDENISLFLIHLSPYFLLKPAQKCLEWLIHR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title