missing translation for 'onlineSavingsMsg'
Learn More

BAP29 Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30507437 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30507437 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30507437 Supplier Novus Biologicals Supplier No. NBP338246C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BAP29 Polyclonal antibody specifically detects BAP29 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAP29
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Bap29, BAP29DKFZp686M2086, B-cell receptor-associated protein 29, BCR-associated protein 29, FLJ53907
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 125-241 of human BAP29 (NP_061332.2).,, Sequence:, TQLAKELSNKGVLKTQAENTNKAAKKFMEENEKLKRILKSHGKDEECVLEAENKKLVEDQEKLKTELRKTSDALSKAQNDVMEMKMQSERLSKEYDQLLKEHSELQDRLERGNKKRL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55973
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.