missing translation for 'onlineSavingsMsg'
Learn More

BarX1 Antibody (BARX1/2759), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18559322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18559322 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18559322 Supplier Novus Biologicals Supplier No. NBP308670AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BarX1 Monoclonal antibody specifically detects BarX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BarX1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BARX1/2759
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1
Host Species Mouse
Immunogen Recombinant fragment of human BarX1 protein (around aa 155-254) (exact sequence is proprietary) (Uniprot: Q9HU1)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56033
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.