missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bassoon Antibody (SAP7F407), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB120-13249-0.025MG
This item is not returnable.
View return policy
Description
Bassoon Monoclonal specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Bassoon | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
| O88778 | |
| BSN | |
| Recombinant rat Bassoon. | |
| Protein G purified | |
| RUO | |
| 8927 | |
| Rat, Mouse | |
| Purified |
| Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SAP7F407 | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| bassoon (presynaptic cytomatrix protein) | |
| Mouse | |
| 400 kDa | |
| 0.025 mg | |
| Primary | |
| This affinity purified detects an ∼400 kDa protein, corresponding to the apparent molecular mass of Bassoon on SDSPAGE immunoblots, in samples from mouse and rat origins. Additional bands between 97 and 400 kDa may also be detected. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only