missing translation for 'onlineSavingsMsg'
Learn More

Bassoon Antibody (SAP7F407), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications

212.10 € - 764.40 €

Specifications

Antigen Bassoon
Clone SAP7F407
Concentration 1.0 mg/mL
Dilution Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Applications Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18030674
View Documents
Novus Biologicals
NB120-13249-0.025MG
0.025 mg
ONLINE EXCLUSIVE PRICE
225.00 €
212.10 € / 25µg
Save 12.90 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18739923
View Documents
Novus Biologicals
NB120-13249
0.05 mg
ONLINE EXCLUSIVE PRICE
422.00 €
399.00 € / 0.05mg
Save 23.00 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18095664
View Documents
Novus Biologicals
NB120-13249-0.2MG
0.2 mg
ONLINE EXCLUSIVE PRICE
809.00 €
764.40 € / 0.20mg
Save 44.60 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bassoon Monoclonal specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Bassoon
1.0 mg/mL
Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Rat, Mouse
bassoon (presynaptic cytomatrix protein)
BSN
Primary
Store at -20C. Avoid freeze-thaw cycles.
400 kDa
SAP7F407
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Monoclonal
Purified
RUO
O88778
8927
Recombinant rat Bassoon.
Protein G purified
This affinity purified detects an ∼400 kDa protein, corresponding to the apparent molecular mass of Bassoon on SDSPAGE immunoblots, in samples from mouse and rat origins. Additional bands between 97 and 400 kDa may also be detected.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.