missing translation for 'onlineSavingsMsg'
Learn More

BCCIP Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30516677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516677 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516677 Supplier Novus Biologicals Supplier No. NBP338567C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BCCIP Polyclonal antibody specifically detects BCCIP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCCIP
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias BCCIPalpha, BRCA2 and CDKN1A interacting protein, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, cdk inhibitor p21 binding protein, P21- and CDK-associated protein 1, Protein TOK-1, TOK-1, TOK-1alpha, TOK1BCCIPbeta, TOK-1beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-314 of human BCCIP (NP_510868.1).,, Sequence:, MASRSKRRAVESGVPQPPDPPVQRDEEEEKEVENEDEDDDDSDKEKDEEDEVIDEEVNIEFEAYSLSDNDYDGIKKLLQQLFLKAPVNTAELTDLLIQQNHIGSVIKQTDVSEDSNDDMDEDEVFGFISLLNLTERKGTQCVEQIQELVLRFCEKNCEKSMVEQLDKFLNDTTKPVGLLLSERFINVPPQIALPMYQQLQKELAGAHRTNKPCGKCYFYLLISKTFVEAGKNNSKKKPSNKKKAALMFANAEEEFFYEKAILKFNYSVQEESDTCLGGKWSFDDVPMTPLRTVMLIPGDKMNEIMDKLKEYLSV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56647
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.