missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Antibody (BL10/411), Novus Biologicals™
Mouse Monoclonal Antibody
260.00 € - 514.00 €
Specifications
| Antigen | Bcl-10 |
|---|---|
| Clone | BL10/411 |
| Concentration | 0.2 mg/ml |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453862
|
Novus Biologicals
NBP2-29422-20ug |
20 ug |
260.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149593
|
Novus Biologicals
NBP2-29422 |
0.1 mg |
514.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-10 Monoclonal specifically detects Bcl-10 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Bcl-10 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| BCL10 | |
| IgG1 κ | |
| Protein A or G purified | |
| BCL10, with an N-terminal caspase recruitment domain (CARD), is found in a number of apoptotic regulatory molecules. It was identified through its direct involvement in t(1;14) of mucosa-associated lymphoid tissue (MALT) lymphoma. Expression of BCL10 was shown to induce NFKB activation in a NIK-dependent pathway. This MAb labels subpopulations of normal B and T cells and is a useful tool for the sub-classification of lymphomas. In MALT lymphomas with the t(1;14) translocation, while 55% of MALT lymphomas lacking this translocation exhibited the same labeling pattern, although at a much lower level. |
| BL10/411 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer | |
| O95999 | |
| 8915 | |
| Human Bcl-10 recombinant protein (epitope aa122-168) (Uniprot: O95999) | |
| Primary | |
| Store at 4C. | |
| 33 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title