missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benazepril Hydrochloride, Mikromol™
Description
Benazepril Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Benazepril hydrochloride |
| CAS | 86541-74-4 |
| Analyte or Component Names | Benazepril hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H28 N2 O5 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benazepril Hydrochloride, (3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride, [S-(R*,R*)]-3-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride, Benazepril hydrochloride, CGS 14824A, CGS 14824A HCl, Lotensin, Lotension |
| InChI Formula | InChI=1S/C24H28N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h3-11,19-20,25H,2,12-16H2,1H3,(H,27,28);1H/t19-,20-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@H]2CCc3ccccc3N(CC(=O)O)C2=O |
| IUPAC Name | 2-[(3S)-3-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetic acid;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?