missing translation for 'onlineSavingsMsg'
Learn More

Benoxaprofen, LoGiCal

Product Code. 30463244
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463244 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463244 Supplier LoGiCal Supplier No. LGCFOR1828.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benoxaprofen
CAS 51234-28-7
Analyte or Component Names Benoxaprofen
Shipping Condition Room Temperature
Molecular Formula C16 H12 Cl N O3
Certifications/Compliance ISO 9001
Synonym (RS)-Benoxaprofen, 2-[2-(4-Chlorophenyl)benzoxazol-5-yl]-propanoic acid, 299582Q1, Benoxaprofen, Compound 90459, Coxigon, LRCL 3794, Lilly 90459, NSC 299582, Opren, Oraflex, Uniprofen, dl-Benoxaprofen, (+/-)-2-(4-Chlorophenyl)-alpha-methyl-5-benzoxazoleacetic acid, (+/-)-Benoxaprofen, 2-(4-Chlorophenyl)-alpha-methyl-5-benzoxazoleacetic acid, 5-Benzoxazoleacetic acid, 2-(4-chlorophenyl)-alpha-methyl-
InChI Formula InChI=1S/C16H12ClNO3/c1-9(16(19)20)11-4-7-14-13(8-11)18-15(21-14)10-2-5-12(17)6-3-10/h2-9H,1H3,(H,19,20)
Solution Type Neat
SMILES CC(C(=O)O)c1ccc2oc(nc2c1)c3ccc(Cl)cc3
IUPAC Name 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 301.72
Quantity 5 mg
Formula Weight 301.0506 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.