missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, Mikromol™

Product Code. 18063660
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063660 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18063660 Supplier Mikromol Supplier No. MM0224.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzalkonium Chloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzalkonium chloride
CAS 8001-54-5
Analyte or Component Names Benzalkonium chloride
Shipping Condition Room Temperature
Molecular Formula C11 H18 N (C2H4)n . Cl
Certifications/Compliance ISO 17034
Synonym Benzalkonium Chloride
Molecular Weight (g/mol) 227.77
Quantity 500 mg
Formula Weight 227.1441 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.