missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,4-diol (Hydroquinone), Mikromol™

Product Code. 17982966
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982966 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982966 Supplier Mikromol Supplier No. MM0632.010250
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzene-1,4-diol (Hydroquinone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroquinone
CAS 123-31-9
Analyte or Component Names Hydroquinone
Shipping Condition Room Temperature
Molecular Formula C6 H6 O2
Certifications/Compliance ISO 17034
Synonym Calcium Dobesilate Monohydrate Imp. A (EP), Dobesilate Imp. A (EP), Hydroquinone, 1,4-Benzoquinol, 1,4-Dihydroxybenzene, 1,4-Phenylenediol, 1,4-p-Benzenediol, 4-Hydroxyphenol, Aida, Arctuvin, BQ(H), Benzohydroquinone, Benzoquinol, Black & White Bleaching Cream, Diak 5, Dihydroquinone, Eldopacque, Eldopaque, Eldopaque Forte, Eldoquin, Eldoquin Forte, HE 5, Hydroquinol, NSC 9247, Phiaquin, Quinol, Solaquin Forte, Solution Q, Tecquinol, Tenox HQ, p-Benzenediol, p-Dihydroquinone, p-Dihydroxybenzene, p-Dioxybenzene, p-Hydroquinone, p-Hydroxyphenol, p-Phenylenediol, p-Quin, Calcium Dobesilate Monohydrate Impurity A, Etamsylate Impurity A, Calcium Dobesilate Impurity A
InChI Formula InChI=1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H
SMILES Oc1ccc(O)cc1
IUPAC Name benzene-1,4-diol
Molecular Weight (g/mol) 110.11
Quantity 250 mg
Formula Weight 110.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.