missing translation for 'onlineSavingsMsg'
Learn More

Benzocaine, Mikromol™

Product Code. 30040606
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040606 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040606 Supplier Mikromol Supplier No. MM0094.000250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzocaine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminobenzoic acid ethyl ester (Benzocaine)
CAS 94-09-7
Analyte or Component Names 4-Aminobenzoic acid ethyl ester (Benzocaine)
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2
Certifications/Compliance ISO 17034
Synonym 4-Aminobenzoic Acid Imp. B (EP), Benzocaine, Benzoic acid, 4-amino-, ethyl ester, Benzoic acid, p-amino-, ethyl ester (8CI), (p-(Ethoxycarbonyl)phenylamine, 4-(Ethoxycarbonyl)aniline, 4-(Ethoxycarbonyl)phenylamine, 4-Aminobenzoic acid ethyl ester, 4-Carbethoxyaniline, Aethoform, Amben ethyl ester, Americaine, Anaesthan-syngala, Anaesthesin, Anaesthin, Anestezin, Anesthesin, Anesthesine, Anesthone, Baby Anbesol, Benzoak, Diet Ayds, Ethoform, Ethyl 4-aminobenzoate, Ethyl aminobenzoate, Ethyl p-aminobenzenecarboxylate, Ethyl p-aminobenzoate, Ethyl p-aminophenylcarboxylate, Gingicaine, Identhesin, Keloform, NSC 41531, NSC 4688, Norcain, Norcaine, Ora-jel, Orabase-B, Orthesin, Parathesin, Parathesine, Slim Mint Gum, Solu H, p-(Ethoxycarbonyl)aniline, p-Aminobenzoic acid ethyl ester, p-Carbethoxyaniline, p-Ethoxycarboxylic aniline, 4-Aminobenzoic Acid Impurity B
InChI Formula InChI=1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3
SMILES CCOC(=O)c1ccc(N)cc1
IUPAC Name ethyl 4-aminobenzoate
Molecular Weight (g/mol) 165.19
Quantity 250 mg
Formula Weight 165.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.