missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Mikromol™

Product Code. 18041790
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18041790 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18041790 Supplier Mikromol Supplier No. MM0278.010250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoic acid
CAS 65-85-0
Analyte or Component Names Benzoic acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O2
Certifications/Compliance ISO 17034
Synonym Benfluorex Hydrochloride Imp. C (EP), Benfluorex Imp. C (EP), Glycopyrronium Bromide Imp. D (EP), Mefenamic Acid Imp. D (EP), Metronidazole Benzoate Imp. C (EP), Benzoic Acid, Benzenecarboxylic acid, Benzeneformic acid, Benzenemethanoic acid, Carboxybenzene, Dracylic acid, E 210, HA 1, HA 1 (acid), MENNO-Florades, NSC 149, Phenylcarboxylic acid, Phenylformic acid, Purox B, Retarder BA, Retardex, Salvo liquid, Solvo powder, Tenn-Plas, VevoVitall, BRL-9421A, Benfluorex Hydrochloride Impurity C, Benzoyl Peroxide Hydrous Impurity B, Glycopyrronium Bromide Impurity D, Mefenamic Acid Impurity D, Metronidazole Benzoate Impurity C, Benfluorex Impurity C, Glycopyrronium Impurity D
InChI Formula InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
SMILES OC(=O)c1ccccc1
IUPAC Name benzoic acid
Molecular Weight (g/mol) 122.12
Quantity 250 mg
Formula Weight 122.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.