missing translation for 'onlineSavingsMsg'
Learn More

BAG3 Antibody (V65P1E8*D2), mFluor Violet 610 SE, R&D Systems™

Product Code. 18609049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609049 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609049 Supplier Novus Biologicals Supplier No. NBP250112MFV610
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

BAG3 Monoclonal specifically detects BAG3 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone V65P1E8*D2
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 9531
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.