missing translation for 'onlineSavingsMsg'
Learn More

Benzoyldiazane (Benzohydrazide), Mikromol™

Product Code. 18060600
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18060600 100 mg 100mg
18021010 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18060600 Supplier Mikromol Supplier No. MM0362.04
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzoyldiazane (Benzohydrazide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoyldiazane (Benzohydrazide)
CAS 613-94-5
Analyte or Component Names Benzoyldiazane (Benzohydrazide)
Shipping Condition Room Temperature
Molecular Formula C7 H8 N2 O
Certifications/Compliance ISO 17025
Synonym Azelastine Hydrochloride Imp. A (EP), Azelastine Imp. A (EP), Benzoyldiazane, Benzohydrazide, Azelastine Hydrochloride Impurity A, Azelastine Impurity A
InChI Formula InChI=1S/C7H8N2O/c8-9-7(10)6-4-2-1-3-5-6/h1-5H,8H2,(H,9,10)
SMILES NNC(=O)c1ccccc1
IUPAC Name benzohydrazide
Molecular Weight (g/mol) 136.15
Quantity 100 mg
Formula Weight 136.0637 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.