missing translation for 'onlineSavingsMsg'
Learn More

Benzyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate (Benzyl Ester of Aceclofenac), Mikromol™

Product Code. 18070580
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070580 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18070580 Supplier Mikromol Supplier No. MM0181.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate (Benzyl Ester of Aceclofenac)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material benzyl[([2-[(2,6-dichlorophenyl)amino]phenyl)acetyl]oxy]acetate
CAS 100499-89-6
Analyte or Component Names benzyl[([2-[(2,6-dichlorophenyl)amino]phenyl)acetyl]oxy]acetate
Shipping Condition Room Temperature
Molecular Formula C23 H19 Cl2 N O4
Certifications/Compliance ISO 17025
Synonym Benzyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate, Aceclofenac Imp. F (EP), Benzyl Ester of Aceclofenac, Aceclofenac Impurity F
InChI Formula InChI=1S/C23H19Cl2NO4/c24-18-10-6-11-19(25)23(18)26-20-12-5-4-9-17(20)13-21(27)30-15-22(28)29-14-16-7-2-1-3-8-16/h1-12,26H,13-15H2
SMILES Clc1cccc(Cl)c1Nc2ccccc2CC(=O)OCC(=O)OCc3ccccc3
IUPAC Name (2-oxo-2-phenylmethoxyethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 444.31
Quantity 25 mg
Formula Weight 443.0691 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.