missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxybenzoate, 99%

Product Code. p-7081667
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11449263

Brand: Thermo Scientific Alfa Aesar A11735.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-18-8 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-18-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016471
InChI Key MOZDKDIOPSPTBH-UHFFFAOYSA-N
Synonym benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
PubChem CID 7180
ChEBI CHEBI:34571
IUPAC Name benzyl 4-hydroxybenzoate
SMILES OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 110°C to 114°C
Quantity 10 g
Beilstein 2115995
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl 4-hydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.