missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyltetradecylammonium Chloride, Mikromol™

Product Code. 17949889
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949889 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949889 Supplier Mikromol Supplier No. MM0224.02
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyldimethyltetradecylammonium Chloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyldimethyltetradecylammonium Chloride
CAS 139-08-2
Analyte or Component Names Benzyldimethyltetradecylammonium Chloride
Shipping Condition Room Temperature
Molecular Formula C23 H42 N . Cl
Certifications/Compliance ISO 17034
Synonym Benzenemethanaminium, N,N-dimethyl-N-tetradecyl-, chloride (1:1), Ammonium, benzyldimethyltetradecyl-, chloride (8CI), Benzenemethanaminium, N,N-dimethyl-N-tetradecyl-, chloride (9CI), Benzyldimethyltetradecylammonium chloride (6CI,7CI), 14BCL, Ammonyx, Arquad DM 14B90, BTC 824P100, Barquat MS 100, Benzyl(tetradecyl)dimethylammonium chloride, Cation M 2-100, Cation M 2-100R, Cyncal, Dibactol, Dimethylbenzylmyristylammonium chloride, Dimethylbenzyltetradecylammonium chloride, JAQ Powdered Quat, M 2-100, M 2-100R, Miristalkonium chloride, Myristalkonium chloride, Myristyl benzyl dimethyl ammonium chloride, Myristyldimethylbenzylammonium chloride, N,N-Dimethyl-N-tetradecylbenzenemethanaminium chloride, N-Benzyl-N,N-dimethylmyristylammonium chloride, N-Benzyl-N,N-dimethyltetradecylammonium chloride, N-Benzyl-N-tetradecyldimethylammonium chloride, N-Tetradecyl-N,N-dimethylbenzylammonium chloride, N-Tetradecyl-N-benzyl-N,N-dimethylammonium chloride, Nissan Cation M 2-100, Nissan Cation M 2-100R, Quarton 14BCl, Querton 14BCL, Sanibond 200LG, Sanisol M 100, TDBAC, TDMBAC, Tetradecylbenzyldimethylammonium chloride, Tetradecyldimethylbenzylammonium chloride, Zeph, Zephiramine, Zephiramine chloride, Zephyramine, Zephyramine (surfactant)
InChI Formula InChI=1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl-dimethyl-tetradecylazanium;chloride
Molecular Weight (g/mol) 368.04
Quantity 500 mg
Formula Weight 367.3006 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.