Learn More
Novus Biologicals™ beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (NK-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30967-200ug
 View more versions of this product
Product Code. 30066805
  • 508.00 € / 200µg
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
NK-1
Western Blot : 1-10 μg/mL, Immunohistochemistry : 2-20 μg/mL, Immunocytochemistry/ Immunofluorescence : 2-20 μg/mL, Immunoprecipitation : 1-2 μg per 100-500 μg of total protein
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Hybridoma produced from mice immunized with human peripheral blood mononuclear cells.
200 μg
Cancer, Immunology
27087
Store at -20°C. Avoid freeze/thaw cycles.
IgM