missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (NK-1), Novus Biologicals™
Mouse Monoclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
|---|---|
| Clone | NK-1 |
| Dilution | Western Blot : 1-10 μg/mL, Immunohistochemistry : 2-20 μg/mL, Immunocytochemistry/ Immunofluorescence : 2-20 μg/mL, Immunoprecipitation : 1-2 μg per 100-500 μg of total protein |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066899
|
Novus Biologicals™
NBP3-30967-50ug |
50 μg |
213.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066805
|
Novus Biologicals™
NBP3-30967-200ug |
200 μg |
508.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry : 2-20 μg/mL, Immunocytochemistry/ Immunofluorescence : 2-20 μg/mL, Immunoprecipitation : 1-2 μg per 100-500 μg of total protein | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
| Hybridoma produced from mice immunized with human peripheral blood mononuclear cells. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| NK-1 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology | |
| Phosphate buffered saline | |
| 27087 | |
| IgM | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title