missing translation for 'onlineSavingsMsg'
Learn More

Beta 2 Adaptin Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30498750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498750 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498750 Supplier Novus Biologicals Supplier No. NBP335911UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Beta 2 Adaptin Polyclonal antibody specifically detects Beta 2 Adaptin in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta 2 Adaptin
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Adapter-related protein complex 2 beta subunit, Adaptor protein complex AP-2 subunit beta, adaptor-related protein complex 2, beta 1 subunit, ADTB2adaptin, beta 2 (beta), AP105B, AP2-BETA, Beta-2-adaptin, Beta-adaptin, CLAPB1AP-2 complex subunit beta, Clathrin assembly protein complex 2 beta large chain, clathrin-associated/assembly/adaptor protein, large, beta 1, DKFZp781K0743, Plasma membrane adaptor HA2/AP2 adaptin beta subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 752-951 of human Beta 2 Adaptin (NP_001025177.1).,, Sequence:, MEMNFTNKALQHMTDFAIQFNKNSFGVIPSTPLAIHTPLMPNQSIDVSLPLNTLGPVMKMEPLNNLQVAVKNNIDVFYFSCLIPLNVLFVEDGKMERQVFLATWKDIPNENELQFQIKECHLNADTVSSKLQNNNVYTIAKRNVEGQDMLYQSLKLTNGIWILAELRIQPGNPNYTLSLKCRAPEVSQYIYQVYDSILKN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.