missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR5 Antibody (85B152.5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NBP1-97728SS
This item is not returnable.
View return policy
Description
TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).
Specifications
| TLR5 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells in 100 ul, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 17570691), Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 24779433), Flow (Intracellular) reported in scientific literature (PMID 24779433) | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| Mouse | |
| 100 kDa | |
| 0.025 mg | |
| Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 7100 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 85B152.5 | |
| Unconjugated | |
| O60602 | |
| TLR5 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction