missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR5 Antibody (85B152.5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 26 publications
213.00 € - 464.00 €
Specifications
| Antigen | TLR5 |
|---|---|
| Clone | 85B152.5 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells in 100 ul, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 17570691), Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 24779433), Flow (Intracellular) reported in scientific literature (PMID 24779433) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017596
|
Novus Biologicals
NBP1-97728SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18786703
|
Novus Biologicals
NBP1-97728 |
0.1 mg |
464.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).Specifications
| TLR5 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| O60602 | |
| 7100 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 85B152.5 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells in 100 ul, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 17570691), Flow (Cell Surface) reported in scientific literature (Wong et al (2007); PMID 24779433), Flow (Intracellular) reported in scientific literature (PMID 24779433) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Canine | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| TLR5 | |
| IgG2a | |
| Protein G purified | |
| 100 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title