missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid Antibody (AB9), Biotin, R&D Systems™

Product Code. 18609046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609046 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18609046 Supplier Novus Biologicals Supplier No. NBP250055B
  • ONLINE EXCLUSIVE PRICE
    651.72 €
    549.15 € / 0.1mL
    Save 102.57 €
    15% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Amyloid
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone AB9
Conjugate Biotin
Formulation PBS
Gene Alias Abeta, Amyloid beta, beta-amyloid peptide
Host Species Mouse
Immunogen This beta Amyloid antibody was developed against amino acids 1-16 of human amyloid Abeta.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.