missing translation for 'onlineSavingsMsg'
Learn More

beta-Catenin Antibody (rCTNNB1/8043), Novus Biologicals™

Product Code. 18651784 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18651784 20 μg 20µg
18659314 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18651784 Supplier Novus Biologicals Supplier No. NBP32370320ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Catenin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rCTNNB1/8043
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Host Species Mouse
Immunogen Recombinant fragment (around aa1-200) of human beta-Catenin protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.