missing translation for 'onlineSavingsMsg'
Learn More

Beta Catenin Mouse, Clone: 12F7, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1499-MSM2-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959857

  • 481.00 € / 100µg
Estimated Shipment: 14-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Beta Catenin
Cell surface and cytoplasmic
12F7
Unconjugated
NP_001895
Mouse
92kDa
Protein A/G Purified
RUO
1499
IgG1 κ
Immunofluorescence
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
CTNNB1
Recombinant fusion protein consisting of the N-terminal half of human beta-Catenin fused to maltose binding protein
HeLa or MCF-7 cells. Breast carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers