missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody (BT7R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18694755 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
Unit Size:
0.03mg
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18694755 0.025 mg 0.03mg
18760504 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18694755 Supplier Novus Biologicals Supplier No. NBP2378300.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone BT7R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:5000, Flow Cytometry 1 μg/test, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunohistochemistry-Paraffin 1:10 - 1:500, Flow (Intracellular), CyTOF-ready
Formulation PBS (pH 7.2) with No Preservative
Gene Alias beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Gene Symbols TUBB
Host Species Mouse
Immunogen KLH conjugated peptide of the N-terminus of Beta-Tubulin.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203068
Test Specificity Detects beta-tubulin in human, monkey, mouse, rat, rabbit and chicken samples with ELISA, Western blot and immunofluorescence applications.
Target Species Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.