missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB100-56459
This item is not returnable.
View return policy
Description
beta Tubulin Polyclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Monkey, Primate, Xenopus samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| beta Tubulin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 203068 | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Monkey, Primate, Xenopus species | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.1 - 1 ug/mL, Simple Western 1:50, Immunohistochemistry 1:100 - 1:1000, Immunohistochemistry-Paraffin 5 - 10 ug/mL | |
| P07437 | |
| TUBB | |
| A synthetic peptide corresponding to a portion of amino acids 400-450 of human b-tubulin was used as immunogen for this antibody, GenBank no. gi 27227551 gb AAN85571.1 . | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen has 100% identity in beta I and beta II tubulin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction