missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications

230.00 € - 517.00 €

Specifications

Antigen beta Tubulin
Concentration 1.0 mg/mL
Dilution Western Blot 0.1 - 1 ug/mL, Simple Western 1:50, Immunohistochemistry 1:100 - 1:1000, Immunohistochemistry-Paraffin 5 - 10 ug/mL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18011774
View Documents
Novus Biologicals
NB100-56459SS
0.025 mg
230.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18775753
View Documents
Novus Biologicals
NB100-56459
0.1 mg
517.00 €
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta Tubulin Polyclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Monkey, Primate, Xenopus samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

beta Tubulin
Western Blot 0.1 - 1 ug/mL, Simple Western 1:50, Immunohistochemistry 1:100 - 1:1000, Immunohistochemistry-Paraffin 5 - 10 ug/mL
Polyclonal
Purified
RUO
PBS with 0.05% Sodium Azide
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
TUBB
IgG
Protein G purified
The amino acid sequence used as immunogen has 100% identity in beta I and beta II tubulin.
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Monkey, Primate, Xenopus species
P07437
203068
A synthetic peptide corresponding to a portion of amino acids 400-450 of human b-tubulin was used as immunogen for this antibody, GenBank no. gi 27227551 gb AAN85571.1 .
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.