missing translation for 'onlineSavingsMsg'
Learn More

Betaine, Mikromol™

Product Code. 17957784
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957784 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17957784 Supplier Mikromol Supplier No. MM3894.001000
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Betaine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betaine (Trimethylglycine)
CAS 107-43-7
Analyte or Component Names Betaine (Trimethylglycine)
Shipping Condition Room Temperature
Molecular Formula C5 H11 N O2
Certifications/Compliance ISO 17025
Synonym Methanaminium, 1-carboxy-N,N,N-trimethyl-, inner salt, Ammonium compounds, substituted, (carboxymethyl)trimethyl-, hydroxide, inner salt (7CI), Betaine (8CI), Methanaminium, 1-carboxy-N,N,N-trimethyl-, hydroxide, inner salt, (Carboxymethyl)trimethylammonium hydroxide inner salt, (Trimethylammonio)acetate, Abromine, Aminocoat, Aquadew AN 100, Auqadew AN 100, Betafin, Betafin BCR, Betafin BP, Betafin BP 20, Bluestim, Cystadane, Fencaijian, FinnStim, Glycine betaine, Glycine, trimethylbetaine, Glycocoll betaine, Glycylbetaine, Greenstim, Loramine AMB 13, Lycine, N,N,N-Trimethylglycine, Oxyneurine, Rubrine C, Trimethylbetaine, Trimethylglycine, Trimethylglycocoll, α-Earleine
InChI Formula InChI=1S/C5H11NO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3
SMILES C[N+](C)(C)CC(=O)[O-]
IUPAC Name 2-(trimethylazaniumyl)acetate
Molecular Weight (g/mol) 117.15
Quantity 1g
Formula Weight 117.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.