missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone Acetate, LoGiCal

Product Code. 30461827
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461827 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461827 Supplier LoGiCal Supplier No. LGCFOR0201.00
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betamethasone acetate
CAS 987-24-6
Analyte or Component Names Betamethasone acetate
Shipping Condition Room Temperature
Molecular Formula C24 H31 F O6
Certifications/Compliance ISO 9001
Synonym Betamethasone Acetate, 9-Fluoro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate, Dexamethasone Acetate Imp. D (EP), Betamethasone 21-acetate, Betamethasone acetate, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,17,21-trihydroxy-16beta-methyl-, 21-acetate (6CI,7CI,8CI), (11beta,16beta)-21-(Acetyloxy)-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, 21-Acetoxy-9alpha-fluoro-11beta,17alpha-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 21-acetate, Dexamethasone Acetate Impurity D
InChI Formula InChI=1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,19-,21-,22-,23-,24-/m0/s1
Solution Type Neat
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 434.5
Quantity 10 mg
Formula Weight 434.2105 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.