missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone Acetate, Mikromol™
Description
Betamethasone Acetate
Specifications
Specifications
| Chemical Name or Material | Betamethasone acetate |
| CAS | 987-24-6 |
| Analyte or Component Names | Betamethasone acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H31 F O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Betamethasone Acetate, 9-Fluoro-11beta,17-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate, Dexamethasone Acetate Imp. D (EP), Betamethasone 21-acetate, Betamethasone acetate, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,17,21-trihydroxy-16beta-methyl-, 21-acetate (6CI,7CI,8CI), (11beta,16beta)-21-(Acetyloxy)-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, 21-Acetoxy-9alpha-fluoro-11beta,17alpha-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 21-acetate, Dexamethasone Acetate Impurity D |
| InChI Formula | InChI=1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,19-,21-,22-,23-,24-/m0/s1 |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?