missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone Sodium Phosphate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Betamethasone Sodium Phosphate
Specifications
Specifications
| Chemical Name or Material | Betamethasone Sodium Phosphate |
| CAS | 151-73-5 |
| Analyte or Component Names | Betamethasone Sodium Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 F O8 P . 2 Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Dexamethasone Sodium Phosphate Imp. B (EP), Betamethasone Sodium Phosphate, (11beta,16beta)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11beta,16beta)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, Bentelan, Betamethasone 21-disodium phosphate, Betamethasone 21-phosphate disodium salt, Betamethasone disodium phosphate, Betamethasone sodium phosphate, Betnesol, Celestan solubile, Disodium betamethasone 21-phosphate, Durabetason, NSC 90616, Steronema, Vista-Methasone, CCI 1517, Dexamethasone Sodium Phosphate Impurity B as Disodium Salt, Dexamethasone Phosphate Impurity B as Disodium Salt |
| InChI Formula | InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15-,16-,17-,19-,20-,21-,22-;;/m0../s1 |
| SMILES | [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-] |
| IUPAC Name | disodium;[2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction