Learn More
Unity Lab Services™ Biosafety Cabinet Ki Test 3rd Party
Product Code. 30286553
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
Product Code. Duration Unit Size
30286553 1 Day Each
1 options
Product Code. 30286553 Supplier Unity Lab Services™ Supplier No. BSCKIMVS
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
KI disc testing for biosafety cabinets, manufactured by third parties such as Labconco and Erlab. This will provide reassurance of cabinet performance and validation of containment efficiency.
  • KI disc testing for biosafety cabinets
  • Manufactured by third parties such as Labconco and Erlab.
  • This will provide reassurance of cabinet performance and validation of containment efficiency.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinets
Duration 1 Day
Product Type Installation, Connection, and Commissioning Service