missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin Antibody (BK-1/39) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50120
This item is not returnable.
View return policy
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for Immunohistochemistry
Specifications
| Biotin | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:10 - 1:500 | |
| Vitamin B4 | |
| Biotin conjugated to keyhole limpet haemocyanin | |
| 0.1 mL | |
| Cancer, Cellular Signaling, Epitope Tags, metabolism | |
| Human | |
| Purified |
| Immunohistochemistry | |
| BK-1/39 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction