missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin Antibody (BK-1/39) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 382.00 €
Specifications
| Antigen | Biotin |
|---|---|
| Clone | BK-1/39 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626126
|
Novus Biologicals
NBP2-50120-0.025ml |
0.025 mL |
220.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627467
|
Novus Biologicals
NBP2-50120 |
0.1 mL |
382.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Biotin | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, Epitope Tags, metabolism | |
| PBS | |
| Biotin conjugated to keyhole limpet haemocyanin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| BK-1/39 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Vitamin B4 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title